missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, Reagent, 99%, Spectrum™ Chemical
IM105, 288-32-4, C3H4N2
Supplier: Spectrum Chemical Mfg Cor IM10512KGBL
Description
Spectrum™ Chemical Imidazole, Reagent is a solid that has no color and is water soluble. It is classified as an diazole and an alkaloid and is an aromatic heterocycle. It belongs to the class of antifungals known as azole and is also used as a corrosion inhibitor on certain metals, notably copper. Imidazole is incorporated into many biological molecules, primarily histidine. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
288-32-4 | |
Poly Pail | |
MFCD00005183 | |
UN3263 | |
N1C=CN=C1 | |
68.08 | |
Reagent |
100% | |
C3H4N2 | |
12 kg | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
1H-imidazole | |
99% |
Chemical Identifiers
288-32-4 | |
68.08 | |
RAXXELZNTBOGNW-UHFFFAOYSA-N | |
N1C=CN=C1 |
C3H4N2 | |
MFCD00005183 | |
1H-imidazole |