missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Reagent, 99%, Spectrum™ Chemical

IM105, 288-32-4, C3H4N2

Supplier:  Spectrum Chemical Mfg Cor IM1055KG

 View more versions of this product

Catalog No. 18603424


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Imidazole, Reagent is a solid that has no color and is water soluble. It is classified as an diazole and an alkaloid and is an aromatic heterocycle. It belongs to the class of antifungals known as azole and is also used as a corrosion inhibitor on certain metals, notably copper. Imidazole is incorporated into many biological molecules, primarily histidine. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
Specifications

Specifications

288-32-4
Poly Pail
MFCD00005183
UN3263
N1C=CN=C1
68.08
Reagent
100%
C3H4N2
5 kg
RAXXELZNTBOGNW-UHFFFAOYSA-N
1H-imidazole
99%

Chemical Identifiers

288-32-4
68.08
RAXXELZNTBOGNW-UHFFFAOYSA-N
N1C=CN=C1
C3H4N2
MFCD00005183
1H-imidazole
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions