missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole Hydrobromide (Low water content) 98.0+%, TCI America™
$154.71 - $462.29
Chemical Identifiers
CAS | 101023-55-6 |
---|---|
Molecular Formula | C3H5BrN2 |
Molecular Weight (g/mol) | 148.99 |
MDL Number | MFCD29765749 |
InChI Key | VWUCIBOKNZGWLX-UHFFFAOYSA-N |
Synonym | Imidazolium Bromide |
PubChem CID | 12203052 |
IUPAC Name | 1H-imidazole hydrobromide |
SMILES | Br.N1C=CN=C1 |
Chemical Identifiers
101023-55-6 | |
148.99 | |
VWUCIBOKNZGWLX-UHFFFAOYSA-N | |
12203052 | |
Br.N1C=CN=C1 |
C3H5BrN2 | |
MFCD29765749 | |
Imidazolium Bromide | |
1H-imidazole hydrobromide |
Specifications
101023-55-6 | |
White-Yellow | |
MFCD29765749 | |
Imidazolium Bromide | |
Br.N1C=CN=C1 | |
148.99 | |
148.99 | |
Crystalline Powder |
221°C | |
C3H5BrN2 | |
1 g | |
VWUCIBOKNZGWLX-UHFFFAOYSA-N | |
1H-imidazole hydrobromide | |
12203052 | |
≥98.0% (HPLC,T) | |
Imidazole Hydrobromide (Low water content) |
Safety and Handling
TSCA : No