missing translation for 'onlineSavingsMsg'
Learn More

Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride, 95%

CAS: 859833-15-1 | C8H6Cl2N2O | 217.049 g/mol

$100.62 - $243.62

Chemical Identifiers

CAS 859833-15-1
Molecular Formula C8H6Cl2N2O
Molecular Weight (g/mol) 217.049
MDL Number MFCD07772809
InChI Key LHLAJCRPSZDOFF-UHFFFAOYSA-N
Synonym imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1
PubChem CID 18525720
IUPAC Name imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride
SMILES C1=CC2=NC=CN2C=C1C(=O)Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH35174MD
View Documents
Thermo Scientific Chemicals
H35174MD
250 mg
Each for $100.62
Only null left
 
AAH3517403
View Documents
Thermo Scientific Chemicals
H3517403
1 g
Each for $243.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

859833-15-1
217.049
LHLAJCRPSZDOFF-UHFFFAOYSA-N
18525720
C1=CC2=NC=CN2C=C1C(=O)Cl.Cl
C8H6Cl2N2O
MFCD07772809
imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1
imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride

Specifications

859833-15-1
C8H6Cl2N2O
250 mg
Hygroscopic
LHLAJCRPSZDOFF-UHFFFAOYSA-N
imidazo[1,2-a]pyridine-6-carbonyl chloride;hydrochloride
18525720
95%
248°C to 250°C
MFCD07772809
UN3261
imidazo 1,2-a pyridine-6-carbonyl chloride hydrochloride, imidazo 1,2-a pyridine-6-carbonylchloride, hydrochloride 1:1, 4-hydroimidazo 1,2-a pyridine-6-carbonyl chloride, chloride, imidazo 1,2-a pyridine-6-carbonyl chloride-hydrogen chloride 1/1
C1=CC2=NC=CN2C=C1C(=O)Cl.Cl
217.049
217.06
Imidazo[1,2-a]pyridine-6-carbonyl chloride hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only