Learn More
Imetit dihydrobromide, 98%
An extremely potent, high affinity agonist at H3 and H4 receptors | CAS: 32385-58-3 | C6H12Br2N4S | 332.06 g/mol
$764.38 - $1214.53
Chemical Identifiers
CAS | 32385-58-3 |
---|---|
Molecular Formula | C6H12Br2N4S |
Molecular Weight (g/mol) | 332.06 |
MDL Number | MFCD00153816 |
InChI Key | DOBOYMKCRRLTRF-UHFFFAOYSA-N |
Synonym | imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide |
PubChem CID | 11957573 |
ChEBI | CHEBI:64151 |
IUPAC Name | 2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide |
SMILES | Br.Br.NC(=N)SCCC1=CN=CN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ62346MF
|
Thermo Scientific Chemicals
J62346MF |
50 mg |
Each for $764.38
|
|
|||||
AAJ62346MC
|
Thermo Scientific Chemicals
J62346MC |
100 mg |
Each for $1,214.53
|
|
|||||
Description
An extremely potent, high affinity agonist at H3 and H4 receptors
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32385-58-3 | |
332.06 | |
DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
11957573 | |
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide |
C6H12Br2N4S | |
MFCD00153816 | |
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
CHEBI:64151 | |
Br.Br.NC(=N)SCCC1=CN=CN1 |
Specifications
32385-58-3 | |
C6H12Br2N4S | |
50 mg | |
Freely soluble in water,practically insoluble in acetone /n | |
Br.Br.NC(=N)SCCC1=CN=CN1 | |
332.06 | |
CHEBI:64151 | |
98% | |
Imetit dihydrobromide |
White | |
MFCD00153816 | |
imetit dihydrobromide, imetit hydrobromide, s-2-imidazol-4-yl ethyl isothiourea, s-2-1h-imidazol-4-yl ethyl isothiourea dihydrobromide, 2-1h-imidazol-4-yl ethyl carbamimidothioate dihydrobromide, s-2-imidazol-4-yl ethyl isothiourea dihydrobromide, imetit dihydrobromide hplc, s-2-imidazol-4-yl ethyl isothiourea 2hbr, s-2-imidazol-4-yl ethyl isothiourea dihydrochloride, 2-2-1h-imidazol-5-yl ethyl isothiourea dihydrobromide | |
DOBOYMKCRRLTRF-UHFFFAOYSA-N | |
2-(1H-imidazol-5-yl)ethyl carbamimidothioate;dihydrobromide | |
11957573 | |
332.06 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only