missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL10 Rabbit anti-Human, Unconjugated, Clone: AOIG-9, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB22029
Description
IL10 Monoclonal antibody specifically detects IL10 in Human samples. It is validated for WB-Ce, IF.Specifications
IL10 | |
Monoclonal | |
Unconjugated | |
IL10 | |
Rabbit | |
100 μg | |
3586 | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot, Immunofluorescence | |
AOIG-9 | |
interleukin 10 | |
CSIF, IL-10, IL10A, MGC126450, MGC126451, TGIF | |
A synthetic peptide corresponding to human IL10 | |
Primary | |
Human | |
Liquid |