Learn More
Ansell HyFlex™ 11-757 Safety Gloves
The HyFlex™ 11-757 Safety Gloves are ultra-thin breathable gloves featuring high level cut protection and touchscreen compatibility.
Supplier:  Ansell 11757110
 View more versions of this product
Catalog No. 17-000-434
Only null left
Explore available promotions
Description
Description
  • Extreme cut protection featuring INTERCEPT™ Technology while maintaining enhanced flexibility and tactility
  • Ultra-lightweight and ultra-thin breathable gloves featuring touchscreen compatibility minimize the need to change or remove gloves
  • High dexterity combined with unmatched comfort and precision while achieving excellent abrasion resistance
  • Tailored fit at the base of the pinky finger better matches natural hand contours
  • Dermatologically approved by Dermatest™
Specifications
Specifications
ANSI ABR 4, ANSI CUT A7
Polyurethane
Specific
Nylon, Stainless Steel Fiber, Polyester, Tungsten
11
11-757
22.2 to 27.4 cm
EN 388:3X31F, EN 420:2003 + A1:2009
Black
Yes
Cut-resistant Gloves
Knit
8.7 to 10.8 in.
Product Suggestions
Product Suggestions