missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine Hydrochloride Solution, USP <251> for Lead, Ricca Chemical
$283.03 - $283.03
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | ClH4NO |
Molecular Weight (g/mol) | 69.49 |
MDL Number | MFCD00051089 |
InChI Key | WTDHULULXKLSOZ-UHFFFAOYSA-N |
Synonym | hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci |
PubChem CID | 443297 |
ChEBI | CHEBI:5807 |
IUPAC Name | hydroxylamine hydrochloride |
SMILES | Cl.NO |
Chemical Identifiers
7732-18-5 | |
69.49 | |
WTDHULULXKLSOZ-UHFFFAOYSA-N | |
443297 | |
hydroxylamine hydrochloride |
ClH4NO | |
MFCD00051089 | |
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
CHEBI:5807 | |
Cl.NO |
Specifications
7732-18-5,67-66-3,5470-11-1,20624-25-3,64-17-5,67-56-1 | |
120 mL | |
MFCD00051089 | |
WTDHULULXKLSOZ-UHFFFAOYSA-N | |
hydroxylamine hydrochloride | |
443297 | |
Laboratory | |
Hydroxylamine hydrochloride solution |
Liquid | |
ClH4NO | |
hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci | |
Cl.NO | |
69.49 | |
CHEBI:5807 | |
Natural Poly Bottle |