missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Titrant T5, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$367.37 - $367.37
Specifications
Specific Gravity | 0.89 |
---|---|
CAS | 7553-56-2,67-56-1 |
CAS Min % | 85,5 |
CAS Max % | 95,15 |
Packaging | Amber Glass Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK2300001C
|
Ricca Chemical Company
RK2300001C |
1 L |
Each for $367.37
|
|
|||||
Specifications
0.89 | |
85,5 | |
Amber Glass Bottle | |
UN1230 |
7553-56-2,67-56-1 | |
95,15 | |
Liquid |