missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Solvent D, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$508.37 - $508.37
Specifications
Specific Gravity | 0.93 |
---|---|
Chemical Name or Material | Solvent D |
CAS | 67-56-1,112-30-1,7446-09-5,288-32-4 |
CAS Min % | 30,30,5,5 |
CAS Max % | 70,70,25,10 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK3100001C
|
Ricca Chemical Company
RK3100001C |
1 L |
Each for $508.37
|
|
|||||
Specifications
0.93 | |
67-56-1,112-30-1,7446-09-5,288-32-4 | |
70,70,25,10 | |
Liquid | |
Karl Fischer |
Solvent D | |
30,30,5,5 | |
Amber Glass Bottle | |
UN1230 |