missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Solvent CX, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$452.00 - $452.00
Specifications
Specific Gravity | 1.04 |
---|---|
Chemical Name or Material | Solvent CX |
CAS | 67-56-1,67-66-3,1330-20-7,7446-09-5,288-32-4 |
CAS Min % | 20,10,10,5,5 |
CAS Max % | 60,40,40,10,10 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK3200001C
|
Ricca Chemical Company
RK3200001C |
1 L |
Each for $452.00
|
|
|||||
Specifications
1.04 | |
67-56-1,67-66-3,1330-20-7,7446-09-5,288-32-4 | |
60,40,40,10,10 | |
Liquid | |
Karl Fischer |
Solvent CX | |
20,10,10,5,5 | |
Amber Glass Bottle | |
UN1992 |