missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Coulometric AG Oven, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$468.47 - $807.57
Specifications
Specific Gravity | 0.97 |
---|---|
Chemical Name or Material | Coulometric AG-Oven |
CAS | 67-56-1,57-55-6,7446-09-5,288-32-4,68007-08-9,111-42-2 |
CAS Min % | 50,20,10,10,10,5 |
CAS Max % | 80,40,30,30,30,10 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK140000500
|
Ricca Chemical Company
RK140000500C |
500 mL |
Each for $468.47
|
|
|||||
RK1400001C
|
Ricca Chemical Company
RK1400001C |
1 L |
Each for $807.57
|
|
|||||
Specifications
0.97 | |
67-56-1,57-55-6,7446-09-5,288-32-4,68007-08-9,111-42-2 | |
80,40,30,30,30,10 | |
Liquid | |
Karl Fischer |
Coulometric AG-Oven | |
50,20,10,10,10,5 | |
Amber Glass Bottle | |
UN1230 |