missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Coulometric AG Oil, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$442.10 - $792.17
Specifications
Specific Gravity | 1.04 |
---|---|
Chemical Name or Material | Coulometric AG-Oil |
CAS | 67-56-1,7446-09-5,67-66-3,288-32-4,1330-20-7,111-42-2 |
CAS Min % | 20,10,10,10,10,10 |
CAS Max % | 60,40,40,30,30,30 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK160000500
|
Ricca Chemical Company
RK160000500C |
500 mL |
Each for $442.10
|
|
|||||
RK1600001C
|
Ricca Chemical Company
RK1600001C |
1 L |
Each for $792.17
|
|
|||||
Specifications
1.04 | |
67-56-1,7446-09-5,67-66-3,288-32-4,1330-20-7,111-42-2 | |
60,40,40,30,30,30 | |
Liquid | |
Karl Fischer |
Coulometric AG-Oil | |
20,10,10,10,10,10 | |
Amber Glass Bottle | |
UN1992 |