missing translation for 'onlineSavingsMsg'
Learn More

HydroSpec™ Composit T5, Ricca Chemical

Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.

$551.63 - $551.63

Specifications

Specific Gravity 1.1
CAS 111-90-0,288-32-4,7446-09-5,7553-56-2
CAS Min % 60,10,5,5
CAS Max % 90,20,20,15
Physical Form Liquid
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RK2000001C
View Documents
Ricca Chemical Company
RK2000001C
1 L
Each for $551.63
Only null left
 
Description

Description

Specifications

Specifications

1.1
60,10,5,5
Liquid
111-90-0,288-32-4,7446-09-5,7553-56-2
90,20,20,15
UN3082
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'