missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Composit K5, Ricca Chemical
Formulated for high performance and reliable results. Karl Fischer reagent for moisture determination.
$595.07 - $595.07
Specifications
Specific Gravity | 1.1 |
---|---|
CAS | 111-90-0,288-32-4,7446-09-5,7553-56-2 |
CAS Min % | 60,10,5,5 |
CAS Max % | 90,20,20,15 |
Packaging | Amber Glass Bottle |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK2200001C
|
Ricca Chemical Company
RK2200001C |
1 L |
Each for $595.07
|
|
|||||
Specifications
1.1 | |
60,10,5,5 | |
Amber Glass Bottle | |
UN3082 |
111-90-0,288-32-4,7446-09-5,7553-56-2 | |
90,20,20,15 | |
Liquid |