missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HydroSpec™ Acid Buffer, Ricca Chemical
Achieve high performance and reliable results. Hydrospec™ supports a wide range of sample types that help you produce the fast, reliable, and reproducible results you are looking for.
$348.19 - $348.19
Specifications
Chemical Name or Material | Acid Buffer |
---|---|
CAS | 67-56-1,288-32-4 |
CAS Min % | 10,90 |
Packaging | Amber Glass Bottle |
Grade | Karl Fischer |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
RK5000001C
|
Ricca Chemical Company
RK5000001C |
1 L |
Each for $348.19
|
|
|||||
Description
- Reliable, Safe, Pyridine-Free
- Available in 500 mL and 1 L sizes
- 1 L Bottles have the Industry Standard GL45 Neck Finish for Convenient Connection
- Stable Formulations and Comprehensive Chemical Compatibility
- 5 Year Shelf Life for Most Reagents
- Technical Support Every Step of the Way
- Tight Specifications for More Accurate Results and Low Lot-to-Lot Variability
- Serving Scientists Successfully for Over 45 Years
Specifications
Acid Buffer | |
10,90 | |
Karl Fischer |
67-56-1,288-32-4 | |
Amber Glass Bottle |