missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroquinone, Crystal, 98.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H108012KG
Specifications
123-31-9 | |
Poly Pail | |
0.985 | |
Crystalline Powder or Lumps |
1 | |
12 kg | |
Reagent |
Chemical Identifiers
123-31-9 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.