missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, 49%, Electronic/Cleanroom Grade, 48.8-49.2%, Spectrum™ Chemical

$2295.07 - $31011.13

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
IUPAC Name hydrogen fluoride
SMILES F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610935
View Documents
Spectrum Chemical Mfg Cor
H10431GL
1 gal. Amber Glass Bottle
Each for $2,295.07
Only null left
 
18610936
View Documents
Spectrum Chemical Mfg Cor
H104355GLBL
55 gal. Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7664-39-3
20.01
KRHYYFGTRYWZRS-UHFFFAOYSA-N
F
FH
MFCD00011346
hydrogen fluoride

Specifications

7664-39-3
Amber Glass Bottle
FH
KRHYYFGTRYWZRS-UHFFFAOYSA-N
hydrogen fluoride
48.8 to 49.2%
1
1 gal.
MFCD00011346
F
20.01
Electronic/Cleanroom Grade
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.