missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., fuming, ≥37%, APHA: ≤10, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., fuming,≥37%, APHA: ≤10
$119.03 - $772.50
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
6000719
|
Honeywell-Fluka
307211LGL |
1 L | Glass Bottle |
Each for $152.28
|
|
||||
6000718
|
Honeywell-Fluka
307211L |
1 L | Plastic Bottle |
Each for $152.25
|
|
||||
6000721
|
Honeywell-Fluka
3072125LGL |
2.5 L | Glass Bottle |
Each for $119.03
|
|
||||
6000720
|
Honeywell-Fluka
3072125L |
2.5 L | Plastic Bottle |
Each for $197.93
|
|
||||
60045845
|
Honeywell-Fluka
307216X1LGL |
6 x 1 L | Glass Bottle |
Case of 6 Each for $772.50
|
|
||||
60045844
|
Honeywell-Fluka
307216X1L |
6 x 1 L | Plastic Bottle |
Case of 6 Each for $772.50
|
|
||||
60045843
|
Honeywell-Fluka
307214X25LGL |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $365.43
|
|
||||
6000722
|
Honeywell-Fluka
307214X25L |
4 x 2.5 L | Plastic Bottle |
Case of 4 Each for $672.20
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
Cl |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 |
Specifications
7647-01-0 | |
Glass Bottle | |
3.23psi (21.1°C), 7.93psi (37.7°C) | |
ClH | |
1789 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
≥37% | |
Hydrochloric Acid, Reag. Ph. Eur. reag. ISO |
1.2g/mL at 25°C | |
≤0.0005% (as SO4) | |
1 L | |
MFCD00011324 MFCD00792839 | |
1098214 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
chlorane | |
313 | |
36.46g/mol | |
ACS/puriss. p.a. |
Safety and Handling
P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314-H335
EINECSNumber : 231-595-7
RTECSNumber : MW4025000