missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
$41.17 - $258.83
Chemical Identifiers
CAS | 7647-01-0 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
LC152201
|
LabChem
LC152201 |
500 mL |
Each for $41.17
|
|
|||||
LC152202
|
LabChem
LC152202 |
1 L |
Each for $43.93
|
|
|||||
LC152204
|
LabChem
LC152204 |
4 L |
Each for $89.43
|
|
|||||
LC152206
|
LabChem
LC152206 |
10 L |
Each for $201.27
|
|
|||||
LC152205
|
LabChem
LC152205 |
20 L |
Each for $258.83
|
|
|||||
Chemical Identifiers
7647-01-0 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
Colorless | |
0.1000N ±0.0005N (0.1M) | |
99.64,0.36 | |
Poly Bottle | |
Traceable to NIST | |
500 mL | |
Liquid | |
MFCD00011324 MFCD00792839 | |
Soluble in water | |
Cl | |
36.46 | |
CHEBI:17883 | |
Certified |
7647-01-0,7732-18-5 | |
1g/mL | |
Passes Test | |
1g/mL | |
Hydrogen chloride | |
ClH | |
HCl | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
36.46 | |
Hydrochloric Acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature