missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 1.00 N, Ricca Chemical
$68.31 - $439.41
Chemical Identifiers
CAS | 7732-18-5 |
---|---|
Molecular Formula | ClH |
Molecular Weight (g/mol) | 36.46 |
MDL Number | MFCD00011324 MFCD00792839 |
InChI Key | VEXZGXHMUGYJMC-UHFFFAOYSA-N |
Synonym | hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof |
PubChem CID | 313 |
ChEBI | CHEBI:17883 |
IUPAC Name | hydrogen chloride |
SMILES | Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
3700-16
|
Ricca Chemical Company
370016 |
500 mL | Natural Poly Bottle |
Each for $68.31
|
|
||||
3700-32
|
Ricca Chemical Company
RI370032 |
1 L | Natural Poly Bottle |
Each for $82.13
|
|
||||
37001CT
|
Ricca Chemical Company
37001CT |
4 L | Cubitainer |
Each for $435.16
|
|
||||
3700-1
|
Ricca Chemical Company
37001 |
4 L | Natural Poly Bottle |
Each for $181.28
|
|
||||
370025
|
Ricca Chemical Company
RI370025 |
10 L | Cubitainer |
Each for $305.59
|
|
||||
3700-5
|
Ricca Chemical Company
RI37005 |
20 L | Cubitainer |
Each for $439.41
|
|
||||
Chemical Identifiers
7732-18-5 | |
36.46 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
313 | |
hydrogen chloride |
ClH | |
MFCD00011324 MFCD00792839 | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
CHEBI:17883 | |
Cl |
Specifications
7732-18-5,7647-01-0 | |
93.0,9.07 | |
500 mL | |
MFCD00011324 MFCD00792839 | |
VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
hydrogen chloride | |
313 | |
Laboratory | |
Hydrochloric Acid |
89.0,8.71 | |
Natural Poly Bottle | |
ClH | |
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
Cl | |
36.46 | |
CHEBI:17883 | |
1.00N |