missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.200 N (N/5) in Isopropyl Alcohol, Ricca Chemical

$144.56 - $600.03

Chemical Identifiers

CAS 67-63-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3802-32
View Documents
Ricca Chemical Company
380232
1 L Natural Poly Bottle
Each for $144.56
Only null left
 
3802-1
View Documents
Ricca Chemical Company
38021
4 L Natural Poly Bottle
Each for $600.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-63-0

Specifications

Colorless
0.200Normal
Isopropyl Alcohol
ClH
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
Laboratory
67-63-0,7732-18-5,7647-01-0
Natural Poly Bottle
1 L
Liquid
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric Acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'