missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic Acid, 48%, 47.0-49.0%, Spectrum™ Chemical
$419.87 - $30400.40
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18610838
|
Spectrum Chemical Mfg Cor
H1029125ML |
125 mL | Amber Glass Bottle |
Each for $419.87
|
|
||||
18610839
|
Spectrum Chemical Mfg Cor
H1029500MLGL |
500 mL | Amber Glass Bottle |
Each for $803.23
|
|
||||
18610835
|
Spectrum Chemical Mfg Cor
H102925LTPV |
2.5 L | Poly-Coated Glass Bottle |
Each for $2,541.07
|
|
||||
18610834
|
Spectrum Chemical Mfg Cor
H10294LTGL |
4 L | Amber Glass Bottle |
Each for $2,804.57
|
|
||||
18610836
|
Spectrum Chemical Mfg Cor
H102920LTBL |
20 L | Poly Drum |
Each for $5,970.93
|
|
||||
18610837
|
Spectrum Chemical Mfg Cor
H1029200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
10035-10-6 |
Specifications
10035-10-6,7732-18-5 | |
Amber Glass Bottle | |
125 mL | |
Reagent |
5e-06 | |
0.0002 | |
47 to 49% |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.