missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine Hydrate Solution, 24 to 26% (iodometric), Honeywell Fluka™

24-26% (iodometric)

$283.80 - $1398.78

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine;hydrate
SMILES NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044908
View Documents
Honeywell Chemicals
156221L
1 L Poly Bottle
Each for $283.80
Only null left
 
60044909
View Documents
Honeywell-Fluka
156226X1L
6 x 1 L Poly Bottle
Case of 6 Each for $1,398.78
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

10217-52-4
32.05
OAKJQQAXSVQMHS-UHFFFAOYSA-N
24654
hydrazine;hydrate
H4N2
MFCD00149931
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
CHEBI:35511
NN

Specifications

Colorless
Liquid
H4N2
MFCD00149931
878137
hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
NN
32.05
CHEBI:35511
24 to 26% (iodometric)
Hydrazine Hydrate Solution
10217-52-4
1 L
NH2NH2 · H2O
UN3293
100°C (212°F)
OAKJQQAXSVQMHS-UHFFFAOYSA-N
hydrazine;hydrate
24654
50.06g/mol
Poly Bottle
Videos
Safety and Handling

Safety and Handling

P201-P260-P280-P304 + P340 + P310-P305 + P351 + P338-P308 + P313

H302-H314-H317-H331-H350-H410

DOTInformation : 6.1/PGIII

SDS
missing translation for 'documents'

missing translation for 'documents'