missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Xylene, Solvent for KF titration, Honeywell Fluka™

Solvent for Karl Fischer titration

$232.88 - $1215.93

Chemical Identifiers

CAS 95-47-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000685
View Documents
Honeywell-Hydranal
378661L
1 L
Each for $232.88
Only null left
 
60046672
View Documents
Honeywell Chemicals
378666X1L
6 x 1 L
Case of 6 Each for $1,215.93
Only null left
 
Description

Description

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
Specifications

Chemical Identifiers

95-47-6

Specifications

31°C (87.8°F)
95-47-6
UN1307
1 L
Glass bottle
HYDRANAL™-Xylene
Videos
Safety and Handling

Safety and Handling

P210-P261-P280-P301 + P310-P305 + P351 + P338-P370 + P378

RTECSNumber : ZE2450000

SDS
missing translation for 'documents'

missing translation for 'documents'