missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Water-in-methanol 5.0, Honeywell Fluka™

Titrating agent for volumetric back titration according to Karl Fischer, 5.00 mg/mL ± 0.05 mg/mL water in methanol

$125.28 - $171.23

Chemical Identifiers

CAS 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name methanol
SMILES CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000974
View Documents
Honeywell Chemicals
34802500ML
500 mL
Each for $125.28
Only null left
 
6000973
View Documents
Honeywell Chemicals
348021L
1 L
Each for $171.23
Only null left
 
Description

Description

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
Specifications

Chemical Identifiers

7732-18-5
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
CHEBI:15377
CO
CH4O
MFCD00004595
962
methanol

Specifications

9°C (48.2°F)
7732-18-5
MFCD00004595
3662861
CO
32.04
CHEBI:15377
HYDRANAL™-Water-in-methanol 5.0
500 mL
CH4O
UN1230
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
962
32.04g/mol
Videos
Safety and Handling

Safety and Handling

P210-P260-P280-P301 + P310-P311

RTECSNumber : PC1400000

SDS
missing translation for 'documents'

missing translation for 'documents'