Learn More
Smart Chemical HYDRANAL™ - Titrant 5, Honeywell Fluka™
Titrating agent for volumetric two-component Karl Fischer titration (methanol-based).
Supplier:  Honeywell Inc 348011LSC
Catalog No. 60-048-621
Only null left
Explore available promotions
Description
Description
Honeywell's Smart Chemical Hydranal™ bottles are embedded with an RFID chip, visible in the form of a new label, that captures essential information for every titration, including article code, product name, lot number, shelf life and numeric concentration.
  • Secure Data Transfer: Ensure correct and complete reagent data every time with traceable information flow.
  • Efficient System Setup: Reduce system set-up time with automated reagent data transfer.
The RFID label is currently compatible with Mettler Toledo's Karl Fischer Compact and Excellence line titrators (when upgraded with external RFID reader) and is available on our best-selling Hydranal Volumetric Titrants.
Specifications
Specifications
11°C
HYDRANAL™-Titrant 5
67-56-1,7553-56-2
UN1230
1 L
0.85 g/mL
Mettler-Toledo Smart Chemical Bottle
Product Suggestions
Product Suggestions