Learn More
HYDRANAL™ - Standard sodium tartrate dihydrate, Standard for Karl Fischer titration (water content ∼15.66%), Honeywell™ Fluka™
Standard for Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)
$130.93 - $130.93
Identifiants chimiques
CAS 6106-24-7
voir les spécifications
Produits 1
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
6002551
Afficher les documents
Honeywell-Hydranal
3469625G
25 g
chaque for $130.93
Il en reste null
Description
Description
Advantages of HYDRANAL-Water Standards:
  • Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty
Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Long shelf life (up to five years)
  • Convenient packaging
  • Supplied with detailed instruction for use
Spécifications
Identifiants chimiques
6106-24-7
Spécifications
25 g
6106-24-7
NONH for all modes of transport
HYDRANAL™-Standard sodium tartrate dihydrate
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'