missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - LipoSolver CM, Reagent for volumetric one-component KF titration in non-polar substances, fats and oils (working medium), Honeywell Fluka™

Medium for volumetric one-component Karl Fischer titration in non-polar substances, fats and oils (chloroform-methanol-based)

$309.15 - $1222.25

Specifications

CAS 67-56-1
MDL Number MFCD02687786
UN Number UN1992
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6000656
View Documents
Honeywell Chemicals
378551L
1 L
Each for $309.15
Case of 6 Each for $1,854.90
Only null left
 
60046666
View Documents
Honeywell-Fluka
378556X1L
6 x 1 L
Case of 6 Each for $1,222.25
Only null left
 
Description

Description

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

Specifications

Specifications

67-56-1
MFCD02687786
UN1992
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'