Learn More
Cytiva HyClone™ CDM4MAb™ Media
Supplier:  Cytiva SH3080101
 View more versions of this product
Catalog No. SH3080101
Only null left
Explore available promotions
Description
Description
Increase process yields for the manufacture of monoclonal antibodies for therapeutic use with HyClone™ CDM4MAb™ Media, a chemically-defined media containing no animal derived components.
Specifications
Specifications
Liquid
0.2 μm Sterile-filtered
No Phenol Red, No HEPES
2° to 8°C, away from light
CDM4MAb Liquid Medium
Glutamine, Poloxamer 188, Sodium Bicarbonate
500 mL bottle
With L-Glutamine
Product Suggestions
Product Suggestions