missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ 0.5 N NaOH, Cytiva

Concentrated sodium hydroxide solutions for bioprocessing applications including pH adjustment and chromatography resin care.

Supplier:  Cytiva SH3056606

 View more versions of this product

Catalog No. SH3056606


Only null left
Explore available promotions

Description

Description

HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean chromatography resins.

  • Manufactured in ISO 9001 and ISO 13485 certified facility.
  • Lot-specific certificate of analysis (CoA) for each lot produced.
  • Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
  • 0.1 μm sterile filtered.
  • All unit sizes are filled in single-use, ready-to-use bags.
  • Raw materials are USP/NF and EP compendial grade.
Specifications

Specifications

Sodium Hydroxide
1310-73-2
0.5 M (0.5 N)
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium hydroxide
NF/USP
Clear, Colorless
HNaO
90 L
[OH-].[Na+]
40.00
Single-use Bioprocessing Container

Chemical Identifiers

1310-73-2
40.00
sodium hydroxide
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
[OH-].[Na+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 12 Months

Recommended Storage : 15°C to 30°C

SDS
Documents

Documents

Promotions

Promotions