missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HyClone™ 0.5 N NaOH, Cytiva
Concentrated sodium hydroxide solutions for bioprocessing applications including pH adjustment and chromatography resin care.
Supplier: Cytiva SH3056604
Description
HyClone™ NaOH solutions in single-use bags offer a convenient format to effectively modify pH and properly care for and clean chromatography resins.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.1 μm sterile filtered.
- All unit sizes are filled in single-use, ready-to-use bags.
- Raw materials are USP/NF and EP compendial grade.
Specifications
Clear, Colorless | |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] | |
40.00 |
1310-73-2 | |
0.5 N | |
15 L | |
sodium hydroxide | |
Single-use Bioprocessing Container |
Chemical Identifiers
1310-73-2 | |
40.00 | |
sodium hydroxide |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
[OH-].[Na+] |
Safety and Handling
ShelfLife : 12 months
Recommended Storage : 15 to 30°C