missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ZNF707 Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100482
Description
Highest antigen sequence indentity to the following orthologs: Mouse (32%), Rat (32%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60886 (PA5-60886. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Specifications
Q96C28 | |
Blocking Assay, Control | |
286075 | |
100 μL | |
Zinc finger protein 707; ZNF707 | |
ZNF707 | |
Human | |
His-ABP-tag | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
≥5.0 mg/mL | |
1 M urea, PBS with no preservative; pH 7.4 | |
Human ZNF707 Control Fragment | |
RUO | |
ZNF707 | |
Unconjugated | |
Recombinant | |
QWEEPWVEDRERPEFQAVQRGPRPGARKSADPKRHCDHPAWAHKKTHVRRERAREGSSFRKGFRLDTDDGQ | |
E. coli | |
>80% by SDS-PAGE and Coomassie blue staining |