missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serpin G1 ELISA Kit
ELISA
$872.00 - $4330.00
Specifications
Accession Number | C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1INH; C1-inhibiting factor; C1NH; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH |
---|---|
Assay Range | 2.4 to 600 ng/mL |
Assay Sensitivity | 2.4 ng/mL |
Conjugate | HRP |
Product Type | ELISA |
Description
Human Serpin G1 quantitates human Serpin G1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin G1.
C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 μg/mL.Specifications
C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1IN; C1INH; C1-inhibiting factor; C1NH; complement component 1 inhibitor; complement component 1 inhibitor (angioedema, hereditary); esterase inhibitor; factor XIIa inhibitor; HAE1; HAE2; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; serpin family G member 1; serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serpin peptidase inhibitor, clade G, member 1; SERPING1; XIIaINH | |
2.4 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
SERPING1 | |
<10 | |
HRP | |
Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
C1 inhibitor | |
4 hrs 45 mins |
2.4 to 600 ng/mL | |
HRP | |
Plasma, Serum, Supernatant | |
710 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
1 hrs 20 mins |