missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse/Rat GDF-11 Recombinant Protein, PeproTech® DFS Item

Recombinant Protein

$132.00 - $20454.06

Specifications

Accession Number O95390, Q9Z1W4, Q9Z217
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 10220, 14561, 29454
Name Human/Mouse/Rat GDF-11
View More Specs

Products 8
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12011100UG DFS Item
View Documents
Gibco™
LS12011100UG
100 μg
Each for $1,268.00
Only null left
 
120111MG DFS Item
View Documents
Gibco™
LS120111MG
2 x 500 μg
Each for $6,010.00
Only null left
 
12-011-01M
View Documents
Gibco™
1201101M
2 x 500 μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
1201120UG DFS Item
View Documents
Gibco™
LS1201120UG
20 μg
Each for $318.00
Only null left
 
12011250UG DFS Item
View Documents
Gibco™
12011250UG
250 μg
Each for $2,220.00
Only null left
 
12011500UG DFS Item
View Documents
Gibco™
12011500UG
500 μg
Each for $3,800.00
Only null left
 
1201150UG DFS Item
View Documents
Gibco™
1201150UG
50 μg
Each for $762.00
Only null left
 
120115UG DFS Item
View Documents
Gibco™
LS120115UG
5 μg
Each for $132.00
Only null left
 
Description

Description

120-11-1MG will be provided as 2 x 500 μg (120-11-500UG). Recombinant Human/Murine/Rat GDF-11 is a 25.0 kDa disulfide-linked homodimer containing two 109 amino acid polypeptide chains. It is highly homologous to myostatin/GDF-8, sharing 90% amino acid sequence identity. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

GDF11 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development.
Specifications

Specifications

O95390, Q9Z1W4, Q9Z217
protein with no preservative
Human/Mouse/Rat GDF-11
RUO
GDF11
Recombinant
Lyophilized
Functional Assay
10220, 14561, 29454
E. coli
BMP11; BMP-11; bone morphogenetic protein 11; GDF11; GDF-11; growth differentiation factor 11; growth/differentiation factor 11
Unconjugated
NLGLDCDEHS SESRCCRYPL TVDFEAFGWD WIIAPKRYKA NYCSGQCEYM FMQKYPHTHL VQQANPRGSA GPCCTPTKMS PINMLYFNDK QQIIYGKIPG MVVDRCGCS
≥ 98% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'