missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human LHX2 Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP105675

Catalog No. RP105675


Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64870 (PA5-64870. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.
Specifications

Specifications

P50458
Blocking Assay, Control
9355
100 μL
ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390
Lhx2
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human LHX2 Control Fragment
RUO
LHX2
Unconjugated
Recombinant
ARDLVYHLNCFTCTTCNKMLTTGDHFGMKDSLVYCRLHFEALLQGEYPAHFNHADVAAAAAAAAAAKSAGLGSAGANPLGLPYYNGVGTVQKGRPRKRKSPGPGADLAAYN
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions