missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-4, Animal-Free Recombinant Protein, PeproTech®

Recombinant Protein

$363.00 - $17369.19

Specifications

Accession Number P08620
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2249
Name Human FGF-4, Animal-Free
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF10031100 DFS Item
View Documents
Gibco™
AF10031100UG
100 μg
Each for $1,112.00
Only null left
 
AF1003101M
View Documents
Gibco™
AF1003101M
2 x 500 μg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AF1003125UG DFS Item
View Documents
Gibco™
AF1003125UG
25 μg
Each for $363.00
Only null left
 
AF10031500 DFS Item
View Documents
Gibco™
AF10031500UG
500 μg
Each for $3,215.00
Only null left
 
Description

Description

AF-100-31-1MG will be provided as 2 x 500 μg (AF-100-31-500UG). Recombinant Human FGF-4 is a 19.7 kDa protein consisting of 182 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Fibroblast growth factor 4 (FGF4) is a member of the fibroblast growth factor (FGF) family that possess broad mitogenic and cell survival activities and play key roles in growth and survival of stem cells during embryogenesis, tissue regeneration, and carcinogenesis. FGF4 was identified by its strong oncogenic transforming activity and is a potent angiogenic factor, expressed in several highly vascularized tumors and also in adult mouse testis, intestine, and brain. Studies on the mouse homolog suggests a function in bone morphogenesis and limb development through the sonic hedgehog (SHH) signaling pathway. Furthermore, FGF4 regulates neural progenitor cell proliferation and neuronal differentiation. Recent studies show a growth-promoting role for FGF4 in human embryonic stem cells and a putative feedback inhibition mechanism by a novel FGF4 splice isoform that may serve to promote differentiation at a later stages of development.
Specifications

Specifications

P08620
protein with no preservative
Human FGF-4, Animal-Free
RUO
Fgf4
Recombinant
Lyophilized
Functional Assay
2249
E. coli
FGF; FGF4; Fgf-4; Fgfk; Fibroblast growth factor; fibroblast growth factor 4; fibroblast growth factor 4 splice isoform; HBGF-4; heparin secretory transforming protein 1; heparin secretory-transforming protein 1; heparin-binding growth factor 1; heparin-binding growth factor 4; HST; Hst1; hst-1; HSTF1; HSTF-1; human stomach cancer, transforming factor from FGF-related oncogene; kaposi sarcoma oncogene; Kfgf; K-FGF; K-fibroblast growth factor; KS3; oncogene HST; transforming protein KS3
Unconjugated
GRGGAAAPTA PNGTLEAELE RRWESLVALS LARLPVAAQP KEAAVQSGAG DYLLGIKRLR RLYCNVGIGF HLQALPDGRI GGAHADTRDS LLELSPVERG VVSIFGVASR FFVAMSSKGK LYGSPFFTDE CTFKEILLPN NYNAYESYKY PGMFIALSKN GKTKKGNRVS PTMKVTHFLP RL
≥ 95% by SDS-PAGE gel and HPLC analyses.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'