missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HTR2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533305
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Monkey (100%) Marmoset, Bat (95%) Horse (90%).
HTR2B is a g-protein coupled receptor for 5-hydroxytryptamine (serotonin). It functions as a receptor for various ergot alkaloid derivatives and psychoactive substances. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors. HTR2B also plays a role in perception of pain, regulation of behavior. It is required to normal proliferation of embryonic cardiac myocytes and normal heart development.Specifications
HTR2B | |
Polyclonal | |
Unconjugated | |
HTR2B | |
5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P41595 | |
HTR2B | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human 5HT2B Receptor. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |