missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HPTLC (High Performance TLC) Glass Plates, Silica Gel 60 Matrix, Binder Polymeric, 200 μm

HPTLC plates may be used to perform high performance thin layer chromatography.

$370.67 - $1330.06

Specifications

Material Glass
Particle Size 10 to 12 μm
Pore Size 60 Å
Fluorescent Indicator No
For Use With (Application) HPTLC, TLC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111009747
View Documents
MilliporeSigma™ Supelco™
Z74022225EA
25/Pk.
Each for $370.67
Only null left
 
111008333
View Documents
MilliporeSigma™ Supelco™
Z74022150EA
50/Pk.
Each for $1,330.06
Only null left
 
Description

Description

Specifications

Specifications

Glass
60 Å
HPTLC, TLC
200 μm
10 to 12 μm
No
Silica Gel 60 Matrix
10 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'