missing translation for 'onlineSavingsMsg'
Learn More

HPCAL1 Monoclonal Antibody (K52064_13F1), Invitrogen™

Mouse Monoclonal Antibody

$1421.98 - $2134.69

Specifications

Antigen HPCAL1
Clone K52064_13F1
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555620
View Documents
Invitrogen™
MA555620
50 μg
Each for $1,421.98
Only null left
 
MA556000
View Documents
Invitrogen™
MA556000
200 μg
Each for $2,134.69
Only null left
 
Description

Description

Sequence of this protein is as follows: MGKQNSKLRP EVLQDLRENT EFTDHELQEW YKGFLKDCPT GHLTVDEFKK IYANFFPYGD ASKFAEHVFR TFDTNGDGTI DFREFIIALS VTSRGKLEQK LKWAFSMYDL DGNGYISRSE MLEIVQAIYK MVSSVMKMPE DESTPEKRTD KIFRQMDTNN DGKLSLEEFI RGAKSDPSIV RLLQCDPSSA SQF

HPCAL1 Monoclonal antibody specifically detects HPCAL1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

HPCAL1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
BDR1; Calcium-binding protein BDR-1; hippocalcin like 1; hippocalcin-like 1; hippocalcin-like 1; visinin like 3; neural visinin-like 3; neural visinin-like protein 3; hippocalcin-like protein 1; HLP2; Hpcal1; neural visinin-like 3; neural visinin-like Ca2+-binding protein type 3; neural visinin-like protein 3; NVL-3; Nvp3; NVP-3; VILIP3; VILIP-3; visinin like 3; visinin-like protein 3; Vnsl3; Vsnl3
Recombinant human HPCAL1 (immunogen range is 1-193).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
HPCAL1
K52064_13F1
Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
P37235, P62748, P62749
3241, 50871, 53602
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'