missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-FlowTM Plus membrane 90 Sheets
Lateral flow test sheets with a nominal capillary flow time of 90 s/4 cm.
$235.42 - $235.42
Specifications
Filter Type | Membrane Filter |
---|---|
Format | Sheet |
Material | Nitrocellulose |
Sterility | Non-sterile |
Flow Rate | 90 sec./4 cm |
Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thickness (English) | Price | Quantity | |||||
HF09002XSS
|
MilliporeSigma
HF09002XSS |
2 mil |
Pack of 5 Each for $235.42
|
|
|||||
HF09004XSS
|
MilliporeSigma
HF09004XSS |
4 mil |
Pack of 5 Each for $235.42
|
|
|||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Contains 5 sample sheets of backed nitrocellulose membrane with a nominal capillary flow time of 90 s/4 cm
- Dimensions: 30.5 x 25 cm
Specifications
Membrane Filter | |
Nitrocellulose | |
90 sec./4 cm | |
Lateral Flow Membrane |
Sheet | |
Non-sterile | |
5 | |
30.5 x 25 cm |