missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma Hi-Flow™ Plus membrane 90 Cards
Lateral flow test cards with a nominal capillary flow time of 90 s/4 cm.
$291.91 - $765.11
Specifications
Filter Type | Membrane Filter |
---|---|
Format | Card |
Material | Nitrocellulose |
Sterility | Non-sterile |
Flow Rate | 90 sec./4 cm |
Catalog Number | Mfr. No. | No. per Case | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. per Case | Price | Quantity | |||||
HF090MC100
|
MilliporeSigma
HF090MC100 |
100/Cs. |
Pack of 100 Each for $765.11
|
|
|||||
HF090MC5PK
|
MilliporeSigma
HF090MC5PK |
5/Cs. |
Pack of 5 Each for $291.91
|
|
|||||
Description
- Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
- Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
- Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
- Dimensions: 6 x 30.1 cm cards
- Flow rate: 90 s/4 cm
Specifications
Membrane Filter | |
Nitrocellulose | |
90 sec./4 cm | |
Lateral Flow Membrane |
Card | |
Non-sterile | |
2 mil | |
6 x 30.1 cm |