missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma Hi-Flow™ Plus membrane 135 Roll

Lateral flow test roll with a nominal capillary flow time of 135 s/4 cm.

$623.85 - $623.85

Specifications

Filter Type Membrane Filter
Format Roll
Material Nitrocellulose
Sterility Non-sterile
Flow Rate 135 sec./4 cm
View More Specs

Products 2
Catalog Number Mfr. No. Thickness (English) Price Quantity  
Catalog Number Mfr. No. Thickness (English) Price Quantity  
SHF1350225
View Documents
MilliporeSigma
SHF1350225
2 mil
Each for $623.85
Only null left
 
SHF1350425
View Documents
MilliporeSigma
SHF1350425
4 mil
Each for $623.85
Only null left
 
Description

Description

  • Designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance
  • Lateral flow tests facilitate quick and convenient detection/quantification of analytes such as antibodies and antigens present in biological samples, including blood, saliva, and urine.
  • Suitable for tests where speed is critical, the sample is plentiful, and sensitivity is not a concern
  • Contains one 100 m slit roll of backed nitrocellulose membrane with a nominal capillary flow time of 135 seconds/4 cm
  • Width: 25 mm
Specifications

Specifications

Membrane Filter
Nitrocellulose
135 sec./4 cm
Lateral Flow Membrane
25 mm
Roll
Non-sterile
1
100 m
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'