missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexyl Nitrite 95.0+%, TCI America™
$109.98 - $891.02
Chemical Identifiers
CAS | 638-51-7 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.175 |
MDL Number | MFCD06797110 |
InChI Key | SGRWGISGVDVSJV-UHFFFAOYSA-N |
PubChem CID | 6365123 |
IUPAC Name | hexyl nitrite |
SMILES | CCCCCCON=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
H106625ML
|
TCI America
H106625ML |
25 mL |
Each for $109.98
|
|
|||||
H1066500ML
|
TCI America
H1066500ML |
500 mL |
Each for $891.02
|
|
|||||
Chemical Identifiers
638-51-7 | |
131.175 | |
SGRWGISGVDVSJV-UHFFFAOYSA-N | |
hexyl nitrite |
C6H13NO2 | |
MFCD06797110 | |
6365123 | |
CCCCCCON=O |
Specifications
638-51-7 | |
52°C | |
MFCD06797110 | |
1993 | |
CCCCCCON=O | |
131.175 | |
131.18 | |
Liquid |
Yellow | |
C6H13NO2 | |
25 mL | |
SGRWGISGVDVSJV-UHFFFAOYSA-N | |
hexyl nitrite | |
6365123 | |
≥95.0% (GC) | |
Hexyl Nitrite |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator