missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaphenoxycyclotriphosphazene 98.0+%, TCI America™
$379.84 - $1320.95
Chemical Identifiers
CAS | 1184-10-7 |
---|---|
Molecular Formula | C36H30N3O6P3 |
Molecular Weight (g/mol) | 693.572 |
MDL Number | MFCD00183774 |
InChI Key | RNFJDJUURJAICM-UHFFFAOYSA-N |
PubChem CID | 136917 |
IUPAC Name | 2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
SMILES | C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7 |
Chemical Identifiers
1184-10-7 | |
693.572 | |
RNFJDJUURJAICM-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene |
C36H30N3O6P3 | |
MFCD00183774 | |
136917 | |
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7 |
Specifications
1184-10-7 | |
White | |
C36H30N3O6P3 | |
5 g | |
C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7 | |
693.572 | |
693.57 | |
Crystalline Powder |
114°C | |
280°C | |
MFCD00183774 | |
RNFJDJUURJAICM-UHFFFAOYSA-N | |
2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene | |
136917 | |
≥98.0% (HPLC,N) | |
Hexaphenoxycyclotriphosphazene |
Safety and Handling
TSCA : No