missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, ACS Reagent, Reagents

Hexanes, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

$129.30 - $8355.40

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 7
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
01-337-941
View Documents
Labchem Inc
C121390015E
15 gal. Steel Drum
Each for $2,858.07
Only null left
 
01-337-940
View Documents
Labchem Inc
C1213900200L
200 L Steel Drum
Each for $8,355.40
Only null left
 
01-337-939
View Documents
Labchem Inc
C121390020E6
20 L Steel Pail
Each for $995.60
Only null left
 
01-337-938
View Documents
Labchem Inc
C12139004A
4 L Poly Bottle
Each for $413.07
Only null left
 
01-337-937
View Documents
Labchem Inc
C12139004C
4 L Glass Bottle
Each for $413.07
Only null left
 
01-337-936
View Documents
Labchem Inc
C12139004C1
4 L Safety Coated Glass
Each for $435.90
Only null left
 
01-337-935
View Documents
Labchem Inc
C1213900500C
500 mL Glass Bottle
Each for $129.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
CCCCCC
C6H14
MFCD02179311
hexane

Specifications

-95°C
Colorless
15 gal.
Liquid
C6H14
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
ACS
110-54-3
68.7°C
Steel Drum
0.665
MFCD02179311
CCCCCC
86.18
Hexanes, ACS Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'