missing translation for 'onlineSavingsMsg'
Learn More

Hexane, B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane), Honeywell Burdick & Jackson

Supplier:  Honeywell-Burdick and Jackson 2114

 View more versions of this product

Catalog No. 6001071


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Hexane
110-54-3
>60.00%,<40.0000%
68.7°C
−26°C
C6H14
UN1208
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CCCCCC
86.18
CHEBI:29021
>99.9%
B&J Brand™, for GC, for pesticide residue analysis, non-spectrophotometric grade, >99.9% (>60% n-Hexane)
>60.0000%
0.659 - 0.673 g/cm3
4 x 4 L
Glass Bottle
MFCD02179311
1730733
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
Pesticide

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Hexanes

SDS
Documents

Documents

Promotions

Promotions