missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorocyclotriphosphazene 97.0+%, TCI America™
$328.83 - $1147.10
Chemical Identifiers
CAS | 15599-91-4 |
---|---|
Molecular Formula | F6N3P3 |
Molecular Weight (g/mol) | 248.93 |
MDL Number | MFCD00192400 |
InChI Key | DKQPXAWBVGCNHG-UHFFFAOYSA-N |
PubChem CID | 139958 |
IUPAC Name | hexafluoro-1,3,5,2λâµ,4λâµ,6λâµ-triazatriphosphinine |
SMILES | FP1(F)=NP(F)(F)=NP(F)(F)=N1 |
Chemical Identifiers
15599-91-4 | |
248.93 | |
DKQPXAWBVGCNHG-UHFFFAOYSA-N | |
hexafluoro-1,3,5,2λâµ,4λâµ,6λâµ-triazatriphosphinine |
F6N3P3 | |
MFCD00192400 | |
139958 | |
FP1(F)=NP(F)(F)=NP(F)(F)=N1 |
Specifications
15599-91-4 | |
White-Yellow | |
F6N3P3 | |
5 g | |
DKQPXAWBVGCNHG-UHFFFAOYSA-N | |
hexafluoro-1,3,5,2λâµ,4λâµ,6λâµ-triazatriphosphinine | |
139958 | |
≥97.0% (GC) | |
Hexafluorocyclotriphosphazene |
27°C | |
51°C | |
MFCD00192400 | |
1759 | |
FP1(F)=NP(F)(F)=NP(F)(F)=N1 | |
248.93 | |
248.93 | |
Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator