missing translation for 'onlineSavingsMsg'
Learn More

Hexadecane 98.0+%, TCI America™

$37.29 - $186.41

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H006625ML
View Documents
TCI America
H006625ML
25 mL
Each for $37.29
Only null left
 
H0066100ML
View Documents
TCI America
H0066100ML
100 mL
Each for $67.11
Only null left
 
H0066500ML
View Documents
TCI America
H0066500ML
500 mL
Each for $186.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

544-76-3
226.448
DCAYPVUWAIABOU-UHFFFAOYSA-N
11006
hexadecane
C16H34
MFCD00008998
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CHEBI:45296
CCCCCCCCCCCCCCCC

Specifications

544-76-3
287°C
MFCD00008998
n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
CCCCCCCCCCCCCCCC
226.448
CHEBI:45296
≥98.0% (GC)
Hexadecane
Colorless
C16H34
25 mL
DCAYPVUWAIABOU-UHFFFAOYSA-N
hexadecane
11006
226.45
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0010

RTECSNumber : ML9200000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'