missing translation for 'onlineSavingsMsg'
Learn More

Heptane, For pesticide residue analysis, Honeywell Riedel-de Haën™

CHROMASOLV™, for pesticide residue analysis

Supplier:  Honeywell Chemicals 3449525L

 View more versions of this product

Catalog No. 6001027


Only null left
Explore available promotions

Description

Description

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
Specifications

Specifications

Heptane
0.684g/mL at 25°C
0.07
433°F
CH3(CH2)5CH3
83 mmHg (37.7°C), 40 mmHg (20°C)
1730763
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2g/mol
CHROMASOLV™
142-82-5
2.5 L
−4°C (24.8°F)
C7H16
MFCD00009544
UN1206
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
≥99% (GC)

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P261-P273-P301 + P310-P331-P501

RTECSNumber : MI7700000

SDS
Documents

Documents

Promotions

Promotions